Product Details

SNP ID
rs3088115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150625654 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAACTTATACTTGAACTACCAATT[C/T]CCTCAAACTTAACATAAATGTAAGA
Phenotype
MIM: 616960
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIMAP6 PubMed Links

Gene Details

Gene
GIMAP6
Gene Name
GTPase, IMAP family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244071.1 3200 UTR 3 NP_001231000.1
NM_001244072.1 3200 UTR 3 NP_001231001.1
NM_024711.5 3200 UTR 3 NP_078987.3

View Full Product Details