Product Details

SNP ID
rs2424147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18288614 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTTGGTGGCGCTGGTGAGTGAG[A/G]CTCCCGCGGGACCCTTGCCGCAGCC
Phenotype
MIM: 604075
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF133 PubMed Links

Gene Details

Gene
ZNF133
Gene Name
zinc finger protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083330.2 Intron NP_001076799.1
NM_001282995.1 Intron NP_001269924.1
NM_001282996.1 Intron NP_001269925.1
NM_001282997.1 Intron NP_001269926.1
NM_001282998.1 Intron NP_001269927.1
NM_001282999.1 Intron NP_001269928.1
NM_001283000.1 Intron NP_001269929.1
NM_001283001.1 Intron NP_001269930.1
NM_001283002.1 Intron NP_001269931.1
NM_001283003.1 Intron NP_001269932.1
NM_001283004.1 Intron NP_001269933.1
NM_001283005.1 Intron NP_001269934.1
NM_001283006.1 Intron NP_001269935.1
NM_001283007.1 Intron NP_001269936.1
NM_001283008.1 Intron NP_001269937.1
NM_003434.5 Intron NP_003425.2
XM_005260819.1 Intron XP_005260876.1
XM_005260820.4 Intron XP_005260877.1
XM_011529336.1 Intron XP_011527638.1
XM_011529337.1 Intron XP_011527639.1
XM_011529338.2 Intron XP_011527640.1
XM_011529339.1 Intron XP_011527641.1
XM_011529340.1 Intron XP_011527642.1
XM_017028041.1 Intron XP_016883530.1
XM_017028042.1 Intron XP_016883531.1
XM_017028043.1 Intron XP_016883532.1
XM_017028044.1 Intron XP_016883533.1
XM_017028045.1 Intron XP_016883534.1
XM_017028046.1 Intron XP_016883535.1
XM_017028047.1 Intron XP_016883536.1
XM_017028048.1 Intron XP_016883537.1
XM_017028049.1 Intron XP_016883538.1
XM_017028050.1 Intron XP_016883539.1
XM_017028051.1 Intron XP_016883540.1
XM_017028052.1 Intron XP_016883541.1
XM_017028053.1 Intron XP_016883542.1
XM_017028054.1 Intron XP_016883543.1
XM_017028055.1 Intron XP_016883544.1
XM_017028056.1 Intron XP_016883545.1
XM_017028057.1 Intron XP_016883546.1
XM_017028058.1 Intron XP_016883547.1

View Full Product Details