Product Details

SNP ID
rs2424209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18473929 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGATGTCCCTCCGCAGCTATAGTT[C/T]CTCTTCTACCCGCTTGAAGAACTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DZANK1 PubMed Links

Gene Details

Gene
DZANK1
Gene Name
double zinc ribbon and ankyrin repeat domains 1
There are no transcripts associated with this gene.

Gene
MIR3192
Gene Name
microRNA 3192
There are no transcripts associated with this gene.

Gene
POLR3F
Gene Name
RNA polymerase III subunit F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282526.1 Intron NP_001269455.1
NM_006466.3 Intron NP_006457.2
XM_011529141.1 Intron XP_011527443.1
XM_017027596.1 Intron XP_016883085.1

View Full Product Details