Product Details

SNP ID
rs2509123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:70079991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACCTGTGACTCACAGGACTGGC[C/T]GGAGGAGCTCAGGGATTGCCATTTG
Phenotype
MIM: 610108
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANO1 PubMed Links

Gene Details

Gene
ANO1
Gene Name
anoctamin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018043.5 Intron NP_060513.5
XM_006718602.2 Intron XP_006718665.1
XM_006718604.2 Intron XP_006718667.1
XM_006718605.2 Intron XP_006718668.1
XM_011545121.2 Intron XP_011543423.1
XM_011545123.2 Intron XP_011543425.1
XM_011545124.2 Intron XP_011543426.1
XM_011545125.2 Intron XP_011543427.1
XM_011545126.2 Intron XP_011543428.1
XM_011545127.2 Intron XP_011543429.1
XM_011545128.2 Intron XP_011543430.1
XM_011545129.2 Intron XP_011543431.1
XM_011545131.2 Intron XP_011543433.1
XM_017017956.1 Intron XP_016873445.1
XM_017017957.1 Intron XP_016873446.1
Gene
ANO1-AS2
Gene Name
ANO1 antisense RNA 2 (head to head)
There are no transcripts associated with this gene.

View Full Product Details