Product Details

SNP ID
rs2500340
Assay Type
Functionally tested
NCBI dbSNP Submissions
64
Location
Chr.1:54999070 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGATCTTGCTCTCCCTTGAAGCCT[C/T]GAGTTGCAGCGATTTCAGTGTCTTC
Phenotype
MIM: 606412
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BSND PubMed Links

Gene Details

Gene
BSND
Gene Name
barttin CLCNK type accessory beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_057176.2 127 UTR 5 NP_476517.1
Gene
TMEM61
Gene Name
transmembrane protein 61
There are no transcripts associated with this gene.

View Full Product Details