Product Details

SNP ID
rs2484112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27069509 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGGAAAATGGAACTCTCACACA[C/T]AGCTGGCAGGGCACACCAAAAAAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP12 PubMed Links

Gene Details

Gene
USP12
Gene Name
ubiquitin specific peptidase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182488.3 Intron NP_872294.2
XM_005266282.2 Intron XP_005266339.1
XM_011534956.2 Intron XP_011533258.1
XM_017020416.1 Intron XP_016875905.1

View Full Product Details