Product Details
- SNP ID
-
rs2540897
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:77182980 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAGCACCACCCTCTGTTATCACCT[C/T]ATCCTTGTTCTCTCTCCGGCCCCTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM63C
PubMed Links
Gene Details
- Gene
- TMEM63C
- Gene Name
- transmembrane protein 63C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020431.3 |
|
Intron |
|
|
NP_065164.2 |
View Full Product Details