Product Details

SNP ID
rs933812191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:233719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGCTTCTGTGAGTTTCAGCACC[A/G]CTCGCCCTCACCTTCGTGTGCAGGC
Phenotype
MIM: 600857
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDHA PubMed Links

Gene Details

Gene
SDHA
Gene Name
succinate dehydrogenase complex flavoprotein subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001294332.1 Intron NP_001281261.1
NM_004168.3 Intron NP_004159.2

View Full Product Details