Product Details

SNP ID
rs2581651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:75214087 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATTACAGGGTTTATTATCTCTAC[C/T]TTGATTCCAAACCTGTGGGATTCTT
Phenotype
MIM: 614427
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSHZ1 PubMed Links

Gene Details

Gene
TSHZ1
Gene Name
teashirt zinc finger homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308210.1 Intron NP_001295139.1
NM_005786.5 Intron NP_005777.3
XM_005266641.3 Intron XP_005266698.1
Gene
ZADH2
Gene Name
zinc binding alcohol dehydrogenase domain containing 2
There are no transcripts associated with this gene.

View Full Product Details