Product Details

SNP ID
rs2601128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73739577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAGTCCCCTCTAGGACATGGCC[C/G]TGGCCATCAGCCAGACTCCACCACA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C15orf59 PubMed Links

Gene Details

Gene
C15orf59
Gene Name
chromosome 15 open reading frame 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039614.2 1450 UTR 3 NP_001034703.1
NM_001303254.1 1450 UTR 3 NP_001290183.1
XM_005254369.4 1450 UTR 3 XP_005254426.1

View Full Product Details