Product Details

SNP ID
rs2630331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70882439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGTTGGCAAGCTGGAGAACCAG[C/G]AAAGCAGATGGTATTGTTGCACACC
Phenotype
MIM: 126090 MIM: 603729
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PCBD1 PubMed Links

Gene Details

Gene
PCBD1
Gene Name
pterin-4 alpha-carbinolamine dehydratase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000281.3 516 Intron NP_000272.1
NM_001289797.1 516 Intron NP_001276726.1
NM_001323004.1 516 Missense Mutation TCC,TGC S88C NP_001309933.1
Gene
SGPL1
Gene Name
sphingosine-1-phosphate lyase 1
There are no transcripts associated with this gene.

View Full Product Details