Product Details

SNP ID
rs2686811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100573563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAAGGCATGGAGGCCCCCAGTC[G/T]CTGTCCCCCCACGGCTCCGTCCGAG
Phenotype
MIM: 604019 MIM: 609097
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AGFG2 PubMed Links

Gene Details

Gene
AGFG2
Gene Name
ArfGAP with FG repeats 2
There are no transcripts associated with this gene.

Gene
FBXO24
Gene Name
F-box protein 24
There are no transcripts associated with this gene.

Gene
LRCH4
Gene Name
leucine rich repeats and calponin homology domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289934.1 Intron NP_001276863.1
NM_002319.4 Intron NP_002310.2
Gene
SAP25
Gene Name
Sin3A associated protein 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168682.1 Intron NP_001162153.1
XM_005250096.3 Intron XP_005250153.1
XM_006715822.3 Intron XP_006715885.1
Gene
ZASP
Gene Name
ZO-2 associated speckle protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289933.1 Intron NP_001276862.1

View Full Product Details