Product Details

SNP ID
rs2681689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:141767741 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAAGGGAAACAGAGGACAAGAAT[C/T]ATTTGTAAATATCTCAGCTTTCTCC
Phenotype
MIM: 606987
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRK7 PubMed Links

Gene Details

Gene
GRK7
Gene Name
G protein-coupled receptor kinase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139209.2 Intron NP_631948.1
XM_017005717.1 Intron XP_016861206.1

View Full Product Details