Product Details

SNP ID
rs2723172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112911022 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCCCACAGGAATTTGTTTATACC[G/T]TTGGCATGGAAAATAGCAGGAAATG
Phenotype
MIM: 605510
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IL37 PubMed Links

Gene Details

Gene
IL37
Gene Name
interleukin 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014439.3 2134 Intron NP_055254.2
NM_173202.1 2134 Intron NP_775294.1
NM_173203.1 2134 Intron NP_775295.1
NM_173204.1 2134 Intron NP_775296.1
NM_173205.1 2134 Intron NP_775297.1
XM_011510963.2 2134 UTR 5 XP_011509265.1
XM_011510964.2 2134 UTR 5 XP_011509266.1

View Full Product Details