Product Details

SNP ID
rs2723617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6475297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCCACCTGAGTTCAGCACATAG[C/T]ACAAGATGCTCCAGTGCTTGTGAAG
Phenotype
MIM: 601638 MIM: 607388 MIM: 605493
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARFIP2 PubMed Links

Gene Details

Gene
ARFIP2
Gene Name
ADP ribosylation factor interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242854.1 3093 UTR 3 NP_001229783.1
NM_001242855.1 3093 UTR 3 NP_001229784.1
NM_001242856.1 3093 UTR 3 NP_001229785.1
NM_012402.3 3093 UTR 3 NP_036534.1
XM_005252840.4 3093 UTR 3 XP_005252897.1
Gene
TIMM10B
Gene Name
translocase of inner mitochondrial membrane 10 homolog B (yeast)
There are no transcripts associated with this gene.

Gene
TRIM3
Gene Name
tripartite motif containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001248006.1 3093 Intron NP_001234935.1
NM_001248007.1 3093 Intron NP_001234936.1
NM_006458.3 3093 Intron NP_006449.2
NM_033278.3 3093 Intron NP_150594.2
XM_011519844.1 3093 Intron XP_011518146.1
XM_011519845.2 3093 Intron XP_011518147.1
XM_017017098.1 3093 Intron XP_016872587.1
XM_017017099.1 3093 Intron XP_016872588.1

View Full Product Details