Product Details

SNP ID
rs2767568
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:208022561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGAAAAGTTGTGGGGAAAGGCA[G/T]GTGGGCTCCGGCCCCTAGTCTCATG
Phenotype
MIM: 601054
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLXNA2 PubMed Links

Gene Details

Gene
PLXNA2
Gene Name
plexin A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025179.3 11384 UTR 3 NP_079455.3
XM_005273164.2 11384 UTR 3 XP_005273221.1
XM_005273165.3 11384 Intron XP_005273222.1

View Full Product Details