Product Details

SNP ID
rs2733920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100889526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGTGGGCGGGAGCGGAGGTCCCA[G/T]GCCAGGCACAGCGTCCTCCTAGCGG
Phenotype
MIM: 100740 MIM: 614469 MIM: 611481
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACHE PubMed Links

Gene Details

Gene
ACHE
Gene Name
acetylcholinesterase (Cartwright blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000665.4 193 Intron NP_000656.1
NM_001282449.1 193 Intron NP_001269378.1
NM_001302621.1 193 Intron NP_001289550.1
NM_001302622.1 193 Intron NP_001289551.1
NM_015831.2 193 Intron NP_056646.1
XM_006715995.2 193 Intron XP_006716058.1
XM_011516225.1 193 Intron XP_011514527.1
XM_011516226.1 193 Intron XP_011514528.1
XM_011516228.2 193 Intron XP_011514530.1
XM_011516229.2 193 Intron XP_011514531.1
XM_017012219.1 193 Intron XP_016867708.1
XM_017012220.1 193 Intron XP_016867709.1
Gene
SRRT
Gene Name
serrate, RNA effector molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128852.1 193 Intron NP_001122324.1
NM_001128853.1 193 Intron NP_001122325.1
NM_001128854.1 193 Intron NP_001122326.1
NM_015908.5 193 Intron NP_056992.4
XM_005250405.2 193 Intron XP_005250462.1
XM_005250406.2 193 Intron XP_005250463.1
XM_005250407.2 193 Intron XP_005250464.1
XM_005250408.2 193 Intron XP_005250465.1
XM_017012290.1 193 Intron XP_016867779.1
XM_017012291.1 193 Intron XP_016867780.1
Gene
UFSP1
Gene Name
UFM1 specific peptidase 1 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015072.3 193 UTR 5 NP_001015072.2

View Full Product Details