Product Details

SNP ID
rs2144440
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:1284756 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCGTGGGTTGGCCACATGGCACG[G/A]CGCAGATCCAGACAGGGTGGAGAGG
Phenotype
MIM: 601328
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ACAP3 PubMed Links

Gene Details

Gene
ACAP3
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
There are no transcripts associated with this gene.

Gene
SCNN1D
Gene Name
sodium channel epithelial 1 delta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130413.3 434 Intron NP_001123885.2
XM_011541899.2 434 Intron XP_011540201.1
XM_011541901.2 434 Intron XP_011540203.1
XM_011541902.2 434 Intron XP_011540204.1
XM_011541905.2 434 Intron XP_011540207.1
XM_011541906.2 434 Intron XP_011540208.1
XM_011541908.2 434 Intron XP_011540210.1
XM_011541920.2 434 Intron XP_011540222.1
XM_011541925.2 434 Intron XP_011540227.1
XM_011541929.2 434 UTR 5 XP_011540231.1
XM_011541932.2 434 Intron XP_011540234.1
XM_011541933.2 434 Intron XP_011540235.1
XM_017002037.1 434 Intron XP_016857526.1
XM_017002038.1 434 Intron XP_016857527.1
XM_017002039.1 434 Intron XP_016857528.1
XM_017002040.1 434 Intron XP_016857529.1
XM_017002041.1 434 Intron XP_016857530.1
XM_017002042.1 434 Intron XP_016857531.1
XM_017002043.1 434 Intron XP_016857532.1
XM_017002044.1 434 Intron XP_016857533.1

View Full Product Details