Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001242844.1 | Intron | NP_001229773.1 | |||
NM_001242845.1 | Intron | NP_001229774.1 | |||
NM_001242846.1 | Intron | NP_001229775.1 | |||
NM_001242847.1 | Intron | NP_001229776.1 | |||
NM_001242848.1 | Intron | NP_001229777.1 | |||
NM_001242849.1 | Intron | NP_001229778.1 | |||
NM_001242850.1 | Intron | NP_001229779.1 | |||
NM_001242851.1 | Intron | NP_001229780.1 | |||
NM_001242852.1 | Intron | NP_001229781.1 | |||
NM_030963.3 | Intron | NP_112225.2 | |||
XM_006715571.3 | Intron | XP_006715634.1 | |||
XM_011536161.2 | Intron | XP_011534463.1 | |||
XM_011536162.2 | Intron | XP_011534464.1 | |||
XM_011536163.2 | Intron | XP_011534465.1 | |||
XM_011536164.2 | Intron | XP_011534466.1 | |||
XM_017011336.1 | Intron | XP_016866825.1 | |||
XM_017011337.1 | Intron | XP_016866826.1 | |||
XM_017011338.1 | Intron | XP_016866827.1 | |||
XM_017011339.1 | Intron | XP_016866828.1 | |||
XM_017011340.1 | Intron | XP_016866829.1 | |||
XM_017011341.1 | Intron | XP_016866830.1 | |||
XM_017011342.1 | Intron | XP_016866831.1 | |||
XM_017011343.1 | Intron | XP_016866832.1 |