Product Details

SNP ID
rs2856946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105843630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTAACTGGATGCCGTCCAGGCCC[C/T]AGGGCAGGGGGCCCTAGCCTGTCTG
Phenotype
MIM: 146910 MIM: 147070 MIM: 147010 MIM: 147170 MIM: 147020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGH PubMed Links

Gene Details

Gene
IGH
Gene Name
immunoglobulin heavy locus
There are no transcripts associated with this gene.

Gene
IGHD
Gene Name
immunoglobulin heavy constant delta
There are no transcripts associated with this gene.

Gene
IGHM
Gene Name
immunoglobulin heavy constant mu
There are no transcripts associated with this gene.

View Full Product Details