Product Details

SNP ID
rs2848706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:31447897 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGACAATGATGGGGCTCTTCCCCA[A/G]AACTACAGGGGCTCTGGCCATCTTC
Phenotype
MIM: 169615
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DSG3 PubMed Links

Gene Details

Gene
DSG3
Gene Name
desmoglein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001944.2 129 Missense Mutation AAA,AGA K7R NP_001935.2
XM_011525850.1 129 Missense Mutation AAA,AGA K7R XP_011524152.1

View Full Product Details