Product Details

SNP ID
rs2133651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:124968156 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATGGAGCACTGTGTCTGGGCGGA[A/G]AGCCTCGGCGTGCCAGGTGTGCATC
Phenotype
MIM: 614182
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HEG1 PubMed Links

Gene Details

Gene
HEG1
Gene Name
heart development protein with EGF like domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020733.1 7005 UTR 3 NP_065784.1
XM_005247666.1 7005 UTR 3 XP_005247723.1

View Full Product Details