Product Details

SNP ID
rs2809922
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:52021279 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATAATAAAAGTGCTTTATCAACTA[C/T]GAAGTCATACCAAAATCTTAGCAGT
Phenotype
MIM: 609448
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TXNDC12 PubMed Links

Gene Details

Gene
TXNDC12
Gene Name
thioredoxin domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015913.3 Intron NP_056997.1

View Full Product Details