Product Details

SNP ID
rs2810690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:23979852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGCTTCTCCGTGACCTTCCTCCC[A/C]CTTCCAGCTCGGAGCGGGATGCTGG
Phenotype
MIM: 613324
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPATA13 PubMed Links

Gene Details

Gene
SPATA13
Gene Name
spermatogenesis associated 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166271.2 227 Intron NP_001159743.1
NM_001286792.1 227 UTR 5 NP_001273721.1
NM_001286793.1 227 Intron NP_001273722.1
NM_001286794.1 227 Intron NP_001273723.1
NM_001286795.1 227 Intron NP_001273724.1
NM_153023.3 227 Intron NP_694568.1

View Full Product Details