Product Details

SNP ID
rs2096258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:71890697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAATCGAGTACCATATTGGAAT[A/T]TAATGTAAATAGGTAACATTTATTG
Phenotype
MIM: 606629
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RIMS1 PubMed Links

Gene Details

Gene
RIMS1
Gene Name
regulating synaptic membrane exocytosis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168407.1 Intron NP_001161879.1
NM_001168408.1 Intron NP_001161880.1
NM_001168409.1 Intron NP_001161881.1
NM_001168410.1 Intron NP_001161882.1
NM_001168411.1 Intron NP_001161883.1
NM_014989.5 Intron NP_055804.2
XM_005248688.3 Intron XP_005248745.1
XM_011535604.2 Intron XP_011533906.2
XM_011535611.2 Intron XP_011533913.2
XM_011535615.2 Intron XP_011533917.2
XM_017010516.1 Intron XP_016866005.1
XM_017010517.1 Intron XP_016866006.1
XM_017010518.1 Intron XP_016866007.1
XM_017010519.1 Intron XP_016866008.1
XM_017010520.1 Intron XP_016866009.1
XM_017010521.1 Intron XP_016866010.1
XM_017010522.1 Intron XP_016866011.1
XM_017010523.1 Intron XP_016866012.1
XM_017010524.1 Intron XP_016866013.1
XM_017010525.1 Intron XP_016866014.1
XM_017010526.1 Intron XP_016866015.1
XM_017010527.1 Intron XP_016866016.1
XM_017010528.1 Intron XP_016866017.1
XM_017010529.1 Intron XP_016866018.1
XM_017010530.1 Intron XP_016866019.1
XM_017010531.1 Intron XP_016866020.1
XM_017010532.1 Intron XP_016866021.1
XM_017010533.1 Intron XP_016866022.1
XM_017010534.1 Intron XP_016866023.1
XM_017010535.1 Intron XP_016866024.1
XM_017010536.1 Intron XP_016866025.1
XM_017010537.1 Intron XP_016866026.1
XM_017010538.1 Intron XP_016866027.1
XM_017010539.1 Intron XP_016866028.1
XM_017010540.1 Intron XP_016866029.1
XM_017010541.1 Intron XP_016866030.1
XM_017010542.1 Intron XP_016866031.1
XM_017010543.1 Intron XP_016866032.1
XM_017010544.1 Intron XP_016866033.1
XM_017010545.1 Intron XP_016866034.1
XM_017010546.1 Intron XP_016866035.1
XM_017010547.1 Intron XP_016866036.1
XM_017010548.1 Intron XP_016866037.1
XM_017010549.1 Intron XP_016866038.1
XM_017010550.1 Intron XP_016866039.1
XM_017010551.1 Intron XP_016866040.1
XM_017010552.1 Intron XP_016866041.1
XM_017010553.1 Intron XP_016866042.1
XM_017010554.1 Intron XP_016866043.1
XM_017010555.1 Intron XP_016866044.1
XM_017010556.1 Intron XP_016866045.1
XM_017010557.1 Intron XP_016866046.1
XM_017010558.1 Intron XP_016866047.1

View Full Product Details