Product Details

SNP ID
rs2812289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127229805 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCACCATACCCAGCAGTTTAAATG[A/C]CTCATGTTTAGGGCTTTTCAGAATC
Phenotype
MIM: 614444
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GARNL3 PubMed Links

Gene Details

Gene
GARNL3
Gene Name
GTPase activating Rap/RanGAP domain like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286779.1 Intron NP_001273708.1
NM_032293.4 Intron NP_115669.3
XM_005252267.3 Intron XP_005252324.1
XM_005252268.2 Intron XP_005252325.1
XM_011519086.2 Intron XP_011517388.1
XM_011519087.2 Intron XP_011517389.1
XM_011519089.2 Intron XP_011517391.1
XM_011519090.2 Intron XP_011517392.1
XM_017015202.1 Intron XP_016870691.1
Gene
RALGPS1
Gene Name
Ral GEF with PH domain and SH3 binding motif 1
There are no transcripts associated with this gene.

View Full Product Details