Product Details

SNP ID
rs2812429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:77792919 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGATAGCTCATGGCCAAGAGGAGA[A/G]TCACAGAAAGATGATGGCGCCAACT
Phenotype
MIM: 604090
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLG5 PubMed Links

Gene Details

Gene
DLG5
Gene Name
discs large MAGUK scaffold protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004747.3 Intron NP_004738.3
XM_005270276.4 Intron XP_005270333.1
XM_006718056.3 Intron XP_006718119.1
XM_006718057.3 Intron XP_006718120.1
XM_011540341.2 Intron XP_011538643.1
XM_011540342.1 Intron XP_011538644.1
XM_011540344.2 Intron XP_011538646.1
XM_011540345.1 Intron XP_011538647.1
XM_011540346.2 Intron XP_011538648.1
XM_011540347.2 Intron XP_011538649.1
XM_017016913.1 Intron XP_016872402.1
XM_017016914.1 Intron XP_016872403.1
XM_017016915.1 Intron XP_016872404.1

View Full Product Details