Product Details

SNP ID
rs2098409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:15108183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCAAACTTAATGAATCAGAGG[A/C]TAATTTTTCTTACTGTACCTGGCTC
Phenotype
MIM: 612664
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RERG PubMed Links

Gene Details

Gene
RERG
Gene Name
RAS like estrogen regulated growth inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190726.1 1867 UTR 3 NP_001177655.1
NM_032918.2 1867 UTR 3 NP_116307.1
XM_017020121.1 1867 UTR 3 XP_016875610.1

View Full Product Details