Product Details

SNP ID
rs2092119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42162374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAGACAGAGCTGGACTATGTTA[A/C]TCCTTTCGACCTCTCAGGAACCTGA
Phenotype
MIM: 603107
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TCF20 PubMed Links

Gene Details

Gene
TCF20
Gene Name
transcription factor 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005650.3 Intron NP_005641.1
NM_181492.2 Intron NP_852469.1
XM_005261722.3 Intron XP_005261779.1
XM_006724313.3 Intron XP_006724376.1
XM_011530353.2 Intron XP_011528655.1
XM_011530354.2 Intron XP_011528656.1

View Full Product Details