Product Details

SNP ID
rs2122694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57014549 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAGAAGGGGTAGGATGGCAATCC[C/T]TCTCATTCATTGACCAAGAGGGGCA
Phenotype
MIM: 162330
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAC3 PubMed Links

Gene Details

Gene
TAC3
Gene Name
tachykinin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178054.1 Intron NP_001171525.1
NM_013251.3 Intron NP_037383.1
Gene
ZBTB39
Gene Name
zinc finger and BTB domain containing 39
There are no transcripts associated with this gene.

View Full Product Details