Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286561.1 | Intron | NP_001273490.1 | |||
NM_006185.3 | Intron | NP_006176.2 | |||
XM_006718564.1 | Intron | XP_006718627.1 | |||
XM_011545054.1 | Intron | XP_011543356.1 | |||
XM_011545055.1 | Intron | XP_011543357.1 | |||
XM_011545056.2 | Intron | XP_011543358.1 | |||
XM_011545057.2 | Intron | XP_011543359.1 | |||
XM_011545058.1 | Intron | XP_011543360.1 | |||
XM_011545059.2 | Intron | XP_011543361.1 | |||
XM_011545060.2 | Intron | XP_011543362.1 | |||
XM_011545061.2 | Intron | XP_011543363.1 | |||
XM_011545062.2 | Intron | XP_011543364.1 | |||
XM_011545063.2 | Intron | XP_011543365.1 | |||
XM_011545064.1 | Intron | XP_011543366.1 | |||
XM_011545065.1 | Intron | XP_011543367.1 | |||
XM_011545066.1 | Intron | XP_011543368.1 | |||
XM_017017830.1 | Intron | XP_016873319.1 | |||
XM_017017831.1 | Intron | XP_016873320.1 |