Product Details

SNP ID
rs2852428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114297337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAAAGTTTTTTTTTTTTCTTTT[G/T]GACTTTAAATTTTATTAAAGTTTGA
Phenotype
MIM: 600008
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NNMT PubMed Links

Gene Details

Gene
NNMT
Gene Name
nicotinamide N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006169.2 Intron NP_006160.1

View Full Product Details