Product Details

SNP ID
rs2859541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43520974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTCCCCTTCTTCCATTCCAGGT[A/T]CATCATCTTCTTAGCAGCTCATCAG
Phenotype
MIM: 600178
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MAP1A PubMed Links

Gene Details

Gene
MAP1A
Gene Name
microtubule associated protein 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002373.5 469 UTR 5 NP_002364.5
XM_017022189.1 469 Silent Mutation GTA,GTT V142V XP_016877678.1

View Full Product Details