Product Details

SNP ID
rs2175328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:25191693 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCCCCTGTTTCTGATATAGGAAA[C/T]GAGGTTTGCAGAAGATGAGTTTCTC
Phenotype
MIM: 616904
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOCK5 PubMed Links

Gene Details

Gene
DOCK5
Gene Name
dedicator of cytokinesis 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322810.1 Intron NP_001309739.1
NM_024940.7 Intron NP_079216.4
XM_011544661.1 Intron XP_011542963.1
XM_011544662.1 Intron XP_011542964.1
XM_011544663.2 Intron XP_011542965.1
XM_017013865.1 Intron XP_016869354.1
XM_017013866.1 Intron XP_016869355.1
XM_017013867.1 Intron XP_016869356.1
XM_017013868.1 Intron XP_016869357.1

View Full Product Details