Product Details
- SNP ID
-
rs2163528
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:28431038 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGAAAAGGCCTAGCTCTCTATGA[C/T]ACAAGAATGGGCTAGTCACATCTCA
- Phenotype
-
MIM: 608519
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO16
PubMed Links
Gene Details
- Gene
- FBXO16
- Gene Name
- F-box protein 16
View Full Product Details