Product Details

SNP ID
rs2877400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:59143543 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTAACATTAGCAAGAGACAGTT[C/T]TGAGCTCTGCTTTTCCTACATCAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEC11C PubMed Links

Gene Details

Gene
SEC11C
Gene Name
SEC11 homolog C, signal peptidase complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307941.1 Intron NP_001294870.1
NM_033280.3 Intron NP_150596.1
XM_011526260.2 Intron XP_011524562.1
XM_017026073.1 Intron XP_016881562.1

View Full Product Details