Product Details

SNP ID
rs2191566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44007237 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCAGTTGCTTCCTTTGTCTCTTTT[G/T]TGTTGACTTTGCATTTGGTTTTTGG
Phenotype
MIM: 604086
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928063 PubMed Links

Gene Details

Gene
LOC101928063
Gene Name
uncharacterized LOC101928063
There are no transcripts associated with this gene.

Gene
ZNF155
Gene Name
zinc finger protein 155
There are no transcripts associated with this gene.

Gene
ZNF230
Gene Name
zinc finger protein 230
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006300.3 Intron NP_006291.2

View Full Product Details