Product Details

SNP ID
rs2199618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:8982649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGTTCAGTGAACGTCGATGGGA[C/T]TGAGTGCTGTGGAGGGTGAGTGAGA
Phenotype
MIM: 606525
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRGAP3 PubMed Links

Gene Details

Gene
SRGAP3
Gene Name
SLIT-ROBO Rho GTPase activating protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033117.2 6904 UTR 3 NP_001028289.1
NM_014850.3 6904 UTR 3 NP_055665.1

View Full Product Details