Product Details

SNP ID
rs2075473
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:36111548 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCATCTGCACACGCCCTCCAGCCTC[A/G]GGACCTGTCCGCCCCTACTTCTCTG
Phenotype
MIM: 616442 MIM: 180662 MIM: 601303 MIM: 613583
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OVOL3 PubMed Links

Gene Details

Gene
OVOL3
Gene Name
ovo like zinc finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302757.1 Intron NP_001289686.1
XM_011527251.2 Intron XP_011525553.1
XM_017027190.1 Intron XP_016882679.1
XM_017027191.1 Intron XP_016882680.1
Gene
POLR2I
Gene Name
RNA polymerase II subunit I
There are no transcripts associated with this gene.

Gene
TBCB
Gene Name
tubulin folding cofactor B
There are no transcripts associated with this gene.

Gene
WDR62
Gene Name
WD repeat domain 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083961.1 Intron NP_001077430.1
NM_173636.4 Intron NP_775907.4
XM_005258809.2 Intron XP_005258866.1
XM_011526837.1 Intron XP_011525139.1
XM_011526838.1 Intron XP_011525140.1
XM_011526839.1 Intron XP_011525141.1
XM_011526840.2 Intron XP_011525142.1
XM_011526841.2 Intron XP_011525143.1
XM_011526842.1 Intron XP_011525144.1
XM_011526843.1 Intron XP_011525145.1
XM_011526844.2 Intron XP_011525146.1
XM_017026665.1 Intron XP_016882154.1

View Full Product Details