Product Details

SNP ID
rs2238614
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:2755341 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGCACCCAGGACGGCTCCTGAGCC[A/G]CGGAGGCGTGGGGTGACCGCAGCCG
Phenotype
MIM: 603419
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SGTA PubMed Links

Gene Details

Gene
SGTA
Gene Name
small glutamine rich tetratricopeptide repeat containing alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003021.3 1742 UTR 3 NP_003012.1
XM_011528178.2 1742 UTR 3 XP_011526480.1

View Full Product Details