Product Details

SNP ID
rs2241843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:111121804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTGCTCTGCAGGAGTCCTGCGG[A/C]CTGTGCCATCCGCACGATTCCCTGG
Phenotype
MIM: 603827
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACOXL PubMed Links

Gene Details

Gene
ACOXL
Gene Name
acyl-CoA oxidase-like
There are no transcripts associated with this gene.

Gene
BCL2L11
Gene Name
BCL2 like 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204106.1 Intron NP_001191035.1
NM_001204107.1 Intron NP_001191036.1
NM_001204108.1 Intron NP_001191037.1
NM_001204109.1 Intron NP_001191038.1
NM_001204110.1 Intron NP_001191039.1
NM_001204111.1 Intron NP_001191040.1
NM_001204112.1 Intron NP_001191041.1
NM_001204113.1 Intron NP_001191042.1
NM_006538.4 Intron NP_006529.1
NM_138621.4 Intron NP_619527.1
NM_138622.3 Intron NP_619528.1
NM_138623.3 Intron NP_619529.1
NM_138624.3 Intron NP_619530.1
NM_138625.3 Intron NP_619531.1
NM_138626.3 Intron NP_619532.1
NM_138627.3 Intron NP_619533.1
NM_207002.3 Intron NP_996885.1
NM_207003.2 Intron NP_996886.1
XM_005263550.3 Intron XP_005263607.1
XM_005263551.2 Intron XP_005263608.1
XM_005263552.3 Intron XP_005263609.1
XM_005263553.2 Intron XP_005263610.1
XM_005263554.2 Intron XP_005263611.1
XM_005263555.3 Intron XP_005263612.1
XM_005263556.3 Intron XP_005263613.1
XM_005263557.4 Intron XP_005263614.1
XM_005263559.2 Intron XP_005263616.1
XM_005263561.3 Intron XP_005263618.1
XM_011510461.2 Intron XP_011508763.1
XM_011510464.2 Intron XP_011508766.1
XM_017003101.1 Intron XP_016858590.1
XM_017003102.1 Intron XP_016858591.1
Gene
LOC400997
Gene Name
uncharacterized LOC400997
There are no transcripts associated with this gene.

View Full Product Details