Product Details

SNP ID
rs2242462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74521685 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGGCGTCCTCATGGGTGTTCAAA[C/G]GGCAACATGCCGAACATGTGTGGTG
Phenotype
MIM: 610705
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD300LB PubMed Links

Gene Details

Gene
CD300LB
Gene Name
CD300 molecule like family member b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174892.3 1783 UTR 3 NP_777552.3
XM_005257027.3 1783 Intron XP_005257084.1

View Full Product Details