Product Details

SNP ID
rs2242655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31659672 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTGCTAGAGATAGGCTGGTCCCA[C/G]TTGAGGGAATCCATTCTTTTGCTCC
Phenotype
MIM: 606907 MIM: 142590 MIM: 115441 MIM: 142610
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APOM PubMed Links

Gene Details

Gene
APOM
Gene Name
apolipoprotein M
There are no transcripts associated with this gene.

Gene
BAG6
Gene Name
BCL2 associated athanogene 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098534.1 1101 Intron NP_001092004.1
NM_001199697.1 1101 Intron NP_001186626.1
NM_001199698.1 1101 Intron NP_001186627.1
NM_004639.3 1101 Intron NP_004630.3
NM_080702.2 1101 Intron NP_542433.1
NM_080703.2 1101 Intron NP_542434.1
XM_011514892.2 1101 Intron XP_011513194.1
XM_017011275.1 1101 Intron XP_016866764.1
XM_017011276.1 1101 Intron XP_016866765.1
XM_017011277.1 1101 Intron XP_016866766.1
XM_017011278.1 1101 Intron XP_016866767.1
XM_017011279.1 1101 Intron XP_016866768.1
XM_017011280.1 1101 Intron XP_016866769.1
XM_017011281.1 1101 Intron XP_016866770.1
XM_017011282.1 1101 Intron XP_016866771.1
XM_017011283.1 1101 Intron XP_016866772.1
XM_017011284.1 1101 Intron XP_016866773.1
XM_017011285.1 1101 Intron XP_016866774.1
XM_017011286.1 1101 Intron XP_016866775.1
XM_017011287.1 1101 Intron XP_016866776.1
XM_017011288.1 1101 Intron XP_016866777.1
XM_017011289.1 1101 Intron XP_016866778.1
XM_017011290.1 1101 Intron XP_016866779.1
XM_017011291.1 1101 Intron XP_016866780.1
XM_017011292.1 1101 Intron XP_016866781.1
XM_017011293.1 1101 Intron XP_016866782.1
XM_017011294.1 1101 Intron XP_016866783.1
XM_017011295.1 1101 Intron XP_016866784.1
XM_017011296.1 1101 Intron XP_016866785.1
XM_017011297.1 1101 Intron XP_016866786.1
XM_017011298.1 1101 Intron XP_016866787.1
Gene
C6orf47
Gene Name
chromosome 6 open reading frame 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021184.3 1101 Missense Mutation AAC,AAG N92K NP_067007.3
Gene
CSNK2B
Gene Name
casein kinase 2 beta
There are no transcripts associated with this gene.

Gene
GPANK1
Gene Name
G-patch domain and ankyrin repeats 1
There are no transcripts associated with this gene.

View Full Product Details