Product Details

SNP ID
rs2239537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101155330 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCATGAATAGAAGAGCTACTGGGG[G/T]AAGGAGAAGACTCCAGGATGTGGCC
Phenotype
MIM: 603531 MIM: 602186
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AP1S1 PubMed Links

Gene Details

Gene
AP1S1
Gene Name
adaptor related protein complex 1 sigma 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283.3 Intron NP_001274.1
Gene
MIR4653
Gene Name
microRNA 4653
There are no transcripts associated with this gene.

Gene
VGF
Gene Name
VGF nerve growth factor inducible
There are no transcripts associated with this gene.

View Full Product Details