Product Details

SNP ID
rs2239745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103497399 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGACAGATGGATGGATGGATGCAC[A/G]ATGTGTCAAACAGGTGCTGGTATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB40A PubMed Links

Gene Details

Gene
RAB40A
Gene Name
RAB40A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080879.2 4174 Intron NP_543155.2
XM_005262083.4 4174 UTR 3 XP_005262140.1
XM_011530872.2 4174 UTR 3 XP_011529174.1

View Full Product Details