Product Details

SNP ID
rs2239816
Assay Type
Functionally tested
NCBI dbSNP Submissions
65
Location
Chr.1:172532503 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
AGGATTCTTGGCACGCCCCTTTCTA[C/T]CTACCAATCAACATCTAGCTCAATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUCO PubMed Links

Gene Details

Gene
SUCO
Gene Name
SUN domain containing ossification factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282750.1 155 Intron NP_001269679.1
NM_001282751.1 155 Intron NP_001269680.1
NM_014283.4 155 Intron NP_055098.1
NM_016227.3 155 Missense Mutation CCT,TCT P11S NP_057311.3
XM_006711374.2 155 Intron XP_006711437.1
XM_006711375.2 155 Intron XP_006711438.1
XM_006711376.1 155 Intron XP_006711439.1

View Full Product Details