Product Details

SNP ID
rs2239899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3003394 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAGGAAGGAGGAACAAGTTTGGG[A/G]AGTTGAAAATCACATTTACTTTGGA
Phenotype
MIM: 123859
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CARS PubMed Links

Gene Details

Gene
CARS
Gene Name
cysteinyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014437.2 Intron NP_001014437.1
NM_001194997.1 Intron NP_001181926.1
NM_001751.5 Intron NP_001742.1
NM_139273.3 Intron NP_644802.1
XM_006718341.3 Intron XP_006718404.1
XM_017018389.1 Intron XP_016873878.1
XM_017018390.1 Intron XP_016873879.1
XM_017018391.1 Intron XP_016873880.1
XM_017018392.1 Intron XP_016873881.1

View Full Product Details