Product Details

SNP ID
rs2285926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:19720790 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGGCTTTCTTATATATGGACTGC[A/C]TATATTATTTCATGAGAATCGTAAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM196 PubMed Links

Gene Details

Gene
TMEM196
Gene Name
transmembrane protein 196
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152774.3 1889 UTR 3 NP_689987.3
XM_005249692.4 1889 Intron XP_005249749.2
XM_011515249.2 1889 Intron XP_011513551.1
XM_011515250.2 1889 Intron XP_011513552.1
XM_017011928.1 1889 Intron XP_016867417.1
XM_017011929.1 1889 Intron XP_016867418.1

View Full Product Details