Product Details

SNP ID
rs2286501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:22817166 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCGTTCCTCCAGAGCAAACCTGCCT[C/G]TTTGAGATTATACCTATATATATAA
Phenotype
MIM: 607980
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TOMM7 PubMed Links

Gene Details

Gene
TOMM7
Gene Name
translocase of outer mitochondrial membrane 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019059.3 Intron NP_061932.1

View Full Product Details