Product Details

SNP ID
rs2274192
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:39028622 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
AATAATATCAAACAGTGTTCTAGAG[A/G]GCTACTTAGCATGATTATCTAATTT
Phenotype
MIM: 603847
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFS5 PubMed Links

Gene Details

Gene
NDUFS5
Gene Name
NADH:ubiquinone oxidoreductase subunit S5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184979.1 Intron NP_001171908.1
NM_004552.2 Intron NP_004543.1

View Full Product Details